To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:toll-like receptor adaptor molecule 2
- Clonality:Polyclonal
- Gene ID:Q86XR7
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TICAM2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TICAM2
- Storage buffer:TIRP Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 26 kDa Observed: 27 kDa
- Storage temperature:TIRP antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:TIRP antibody was raised against a peptide corresponding to 16 amino acids at the C-terminus of human TIRP. The immunogen is located within the last 50 amino acids of TIRP.
- Tested applications:Immunocytochemistry
- Purification:TIRP Antibody is Ion exchange chromatography purified.
- Cat. no.:89415-626
- Supplier No.:3206
Specifications
About this item
TIRP Antibody: TIRP is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TIRP, along with other molecules such as TRIF, MAL, and MyD88, serves as an adaptor protein that allows for the interaction and activation of the IL-1R-associated kinase (IRAK) family, the subsequent activation of TNF receptor associated factor (TRAF)-6, and ultimately the activation of NF-kappa B. Expression of TIRP appears to be essential for TLR4 signalling.
Type: Primary
Antigen: TICAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human