To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:conserved helix-loop-helix ubiquitous kinase
- Clonality:Polyclonal
- Gene ID:O15111
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CHUK
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CHUK
- Storage buffer:IKK alpha Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:85 kDa
- Storage temperature:IKK alpha antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:IKK alpha antibody was raised against a 19 amino acid peptide near the carboxy terminus of human IKK alpha. The immunogen is located within the last 50 amino acids of IKK alpha.
- Tested applications:Immunocytochemistry
- Purification:IKK alpha Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89415-284
- Supplier No.:2025
Specifications
About this item
IKK alpha Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFa, and bacteria product LPS. NF-kappa B is associated with I kappa B proteins in the cell cytoplasm, which inhibit NF-kappa B activity. The long-sought I kappa B kinase (IKK), which phosphorylates I kappa B, and mediates I kappa B degradation and NF-kappa B activation, was recently identified by several laboratories. IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta ). IKK alpha and IKK beta interact with each other and both are essential for the NF-kappa B activation. IKK alpha specifically phosphorylates IkB-alpha. IKK alpha is expressed in variety of human tissues.
Type: Primary
Antigen: CHUK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human