To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:mitogen-activated protein kinase kinase kinase 14
- Clonality:Polyclonal
- Gene ID:Q99558
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:MAP3K14
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MAP3K14
- Storage buffer:NIK Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 104 kDa
- Storage temperature:NIK antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:NIK antibody was raised against a 17 amino acid peptide near the carboxy terminus of human NIK. The immunogen is located within the last 50 amino acids of NIK.
- Tested applications:Immunocytochemistry
- Purification:NIK Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89415-232
- Supplier No.:1129
Specifications
About this item
NIK Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNF alpha , LPS and mitogens. A serine/threonine protein kinase which mediates NF-kappa B activation by IL-1, TNF alpha and CD95 was identified recently and designated NIK (for NF-kappa B inducing kinase). NIK is an activator of I kappa B kinase alpha and beta (IKK alpha and IKK beta ). Therefore, NIK is a key molecule in the NF-kappa B signaling pathway leading to the induction of a variety of gene expression in response to proinflammatory cytokines and bacteria products.
Type: Primary
Antigen: MAP3K14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human