Specifications
- Description:Dry Milk Powder, Blocking Agent
- Size:4 x 25 g
- Cat. no.:89410-742
- Supplier No.:22012
Specifications
About this item
Milk is commonly used in blocking buffers minimize background noise in immunodetection procedures such as Western Blotting and immunostaining
Milk can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or used for blocking and dilution of antibodies during immunostaining procedures.