SP Bel-Art Stir & Add™ Cuvette Mixers, Bel-Art Products, a part of SP
Catalog #: 59063-125
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:9.1× 9.1 mm (0.36×0.36”)
- Length:11.4 cm (4½") (Shaft)
- Description:Cuvette Mixer for 10 mm Light Path Cells
- Cat. no.:59063-125
- Supplier No.:377520000
Specifications
About this item
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents.
- Clear acrylic construction
- Straight handle
- Complete mixing with simple up and down motion
Up and down motion in cell provides complete mixing. Shaft diameter is 3.17 mm (0.125").
Customers Who Bought This Also Bought