About this item
SensION+ 9655 is a combination fluoride ion selective electrode (ISE) with an epoxy body, non-refillable, single-junction reference, and non-replaceable crystalline membrane selective to fluoride ions in solution
This electrode requires virtually no maintenance, and it provides fast, stable, and accurate responses in a variety of sample types.
The 9655 has a BNC connector for laboratory use with the Hach sensION+ MM340 GLP pH/ISE Laboratory Meter or the sensION+ MM374 Laboratory Multimeter. The 9655 is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality laboratory applications.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA89501-154
- Range:
- 0.01 mg/L to 19,000 mg/L F-
- Temperature range:
- 5 to 50 °C
- Junction:
- Porous Teflon Annular Ring