SP Bel-Art Water Bath Racks, Bel-Art Products, a part of SP
Supplier: Avantor
Certificates
About this item
Molded from a special blend of polypropylene that is heavier than water, these racks sink to the bottom of the bath and will not tip over.
- Polypropylene blend construction
- Sinks in water
- Will not tip over
- Autoclavable
All sizes are 23Lx110Wmm (8⅜x4⁵⁄₁₆"). Steam autoclavable at 121[°C(250°F).
Customers Who Bought This Also Bought