Specifications
- Description:Ward's® Sealed With A Kiss Lab Activity - Refill
- No. of Lab Groups:—
- Cat. no.:470024-230
- Supplier No.:WARD470024-230
Specifications
About this item
Are lip prints like finger prints?
- NGSS Physical Science DCI PS1
- Pre and post-laboratory assessments
- Links to real world concepts
Someone has been mailing anonymous love letters to the captain of the football team and sealing each one with a kiss. He wants his fellow classmates to help him get to the bottom of this mystery. In this lab activity, students will use paper chromatography to compare the lipstick worn by each female suspect to that of the lipstick found on the envelopes. Students will then analyze the lip prints of each suspect in order to positively identify the “guilty” party. While completing this lab activity, students will employ Framework Science Standard PS1.
This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific “Did You Know” concepts that link the real world with the scientific standards explored. A student copymaster is also included. This kit includes enough materials to be performed once for eight group of students.