SP Bel-Art Pipette Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Designed for the safe and orderly storage of up to 28 pipettes.
- This polypropylene stand has a two-section top plate
- One section has 10 holes, 14 mm in diameter
- Other section has 18 holes, 11 mm in diameter
Holes in the lower plate are 4 mm in diameter and support the tips of the pipettes as they are held in the stand.
Customers Who Bought This Also Bought