Stainless Steel Support Rods, Ace Glass
Supplier: Ace Glass
Certificates
About this item
303 stainless steel support rods, for use in place of aluminum or fiberglass
For use with all standard laboratory connectors.
Customers Who Bought This Also Bought