Labjaws FootPlate, Ace Glass Incorporated
Catalog #: 89084-716
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Labjaws FootPlate for Frame
- Cat. no.:89084-716
- Supplier No.:11174-13
Specifications
About this item
The Labjaws Footplate holds Flexaframe poles
It secures them to floors, walls and tables with screws. Mount lattice permanently.
Customers Who Bought This Also Bought