Labjaws Hook Connectors, Ace Glass
Catalog #: 89084-718
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Labjaws Hook Connectors
- Cat. no.:89084-718
- Supplier No.:11169-24
Specifications
About this item
These Labjaws Hook Connectors lock two Flexaframe rods at a 90° angle
Does not slip. Install without disassembling frame. Supplied with Flexaframe sets.
Customers Who Bought This Also Bought