Labjaws Buret Holder, Ace Glass Incorporated
Catalog #: 89084-682
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:100 ml
- Diameter ×L:13 mm (0.51") x (578 mm (22.76")
- Description:Labjaws Buret Holder, Tallboys by Troemmer
- Cat. no.:89084-682
- Supplier No.:11051-15
Specifications
About this item
This buret support provides a large, stable work area, combining a 612 cm2 glazed porcelain base, for ample work space, with a double buret holder
The base is easy to clean, impervious to all ordinary reagents, and withstands high heat (up to 260°C). Weight is 2.3 Kg. The unit has four rubber feet to prevent sliding. It is supplied with a 60 cm high anodized aluminum support rod.
Customers Who Bought This Also Bought