Conical Flange Reaction Head, 4 Neck, Ace Glass
Catalog #: 89054-980
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Conical Flange Reaction Head, 4 Necks
- Cat. no.:89054-980
- Side ST Joint Size:35/25
- Center Joint:35/25
- Supplier No.:6489-20
Specifications
About this item
Standard 4-inch (100mm) head with three socket joints in line, plus a standard taper 14/35 side joint for thermometer
For use with certain ACE reaction flasks. Fabricated from borosilicate glass
Customers Who Bought This Also Bought