High Vacuum Bearing, Trubore®, Ace Glass Incorporated
Catalog #: 89061-814
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:High Vacuum Bearing, Trubore®, Liquid Seal, High Vacuum Glass, 10 mm
- Cat. no.:89061-814
- ST Joint Size:24/40
- Supplier No.:8055-10
Specifications
About this item
This bearing is the standard liquid seal type except that the use of Trubore® tubing enables this unit to perform very satisfactorily under high vacuum conditions
I.D. is 10 mm. For use with 10 mm shafts.
Customers Who Bought This Also Bought