Multi-Paddle Agitator, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
PTFE bottom agitator for large reactors
Multi-paddle solid assembly fits 28 mm glass shafts. Agitator is attached to the shaft by a PTFE bolt. The bottom of the center piece is bored out to receive the pin from the flush seal valve. This agitator when coupled to the valve greatly reduces flexing or wobble from the shaft.
Ordering information: Shaft separately.
Related Information
VF1013 30 to 50L Jackted Reactors VF1014 75 to 150L Jackted Reactors
Product Literature
Customers Who Bought This Also Bought