Water-Cooled Trubore® Bearing, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Trubore® 19 mm I.D., stirrer bearing with standard taper joint bottom
Jacketed for water cooling. Designed for use with 19 mm O.D. precision ground glass shafts and PTFE shafts with stainless steel core. Hose connections are size D for use with 3/8" I.D. tubing.
Customers Who Bought This Also Bought