Material Introduction Trubore® Bearing, Ace Glass Incorporated
Catalog #: 89061-812
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Length:10 mm
- Description:Material Introduction/Gas Dispersion Trubore® Bearing
- Cat. no.:89061-812
- ST Joint Size:24/40
- Supplier No.:8053-10
Specifications
About this item
Bearing is especially useful in controlled atmosphere work when used with our solid shaft and balancing pressure differentials across the bearing
Material introduction or gaseous dispersions can be obtained using our hollow, precision ground stirring shafts.
Customers Who Bought This Also Bought