Ranger® Count 3000 Counting Scales, Ohaus
Supplier: Ohaus
Certificates
About this item
The Ranger® Count 3000 is a high-precision dedicated counting scale with advanced software for counting and packaging parts
The large bright displays and SmarText™ navigation make the Ranger® Count 3000 extremely simple to use.The three-window backlit LCD display includes a centrally-located count display and separate weight and piece weight displays. Eight function keys, including dedicated SAMPLE and TARE keys, and full numeric keypad make setup and input operations simple and straightforward. All units feature ABS housing and a stainless steel platform.
With low Average Piece Weight (APW) alert and automatic APW recalculation, these units offer 1:1,500,000 internal counting resolution to handle counting even the smallest parts precisely.
Certifications: This product conforms to the EMC Directive 2004/108/EC, and the Low Voltage Directive 2006/95/EC.
Ordering information: Ethernet, USB connectivity, and additional RS232 ports are all available as accessory items.
Customers Who Bought This Also Bought