316 Stainless Steel Stirrer Blades for 10 mm Button Shafts, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
The 316 stainless steel stir blades for 10 mm O.D.
- Button type stir shafts are football shaped
- Holed versions provide even greater agitation
Customers Who Bought This Also Bought