Explorer® Precision Balances, with External Calibration, Ohaus
Supplier: Ohaus
Certificates
About this item
A large color touch-screen display, icon-driven application software that features 14 unique application modes, and an adjustable angle display make Explorer the most advanced balance in its class.
These models feature traditional external calibration in which external weights are used to calibrate the balance to ensure accuracy.
Certifications: This product conforms to the EMC Directive 2004/108/EC, and the Low Voltage Directive 2006/95/EC.
Ordering information: Ethernet, USB connectivity, and additional RS232 ports are all available as accessory items.
Customers Who Bought This Also Bought