Orthophosphoric acid ≥85.0% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Phosphoric acid
Formula:H₃PO₄
MW:98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
MDL Number:MFCD00011340
CAS Number:7664-38-2
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 28 °C
:Flash Pt:
:Flash Pt:
:M.W. 98 g/mol
Storage Temperature:Ambient
EINECS:231-633-2
:158
UN:1805
:8
:III
:GHS05
:12,07500
:14,07344
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (H3PO4):85.0% min.
- Colour (APHA):10 max.
- Insoluble Matter:0.001% min.
- Chloride (Cl):3 ppm max.
- Nitrate (NO3):5 ppm max.
- Sulfate (SO4):0.003% max.
- Volatile acids (as CH3COOH):0.001% max.
- Antimony (Sb):0.002% max.
- Calcium (Ca):0.002% max.
- Magnesium (Mg):0.002% max.
- Potassium (K):0.005% max.
- Sodium (Na):0.025% max.
- Arsenic (As):1 ppm max.
- Heavy metals (as Pb):0.001% max.
- Iron (Fe):0.003% max.
- Manganese (Mn):0.5 ppm max.
- Reducing substances:Passes Test
- Orthophosphoric acid ≥85.0% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_ANAC70012-463.pdf spec_ANAC70012-543.pdf spec_CA71007-998 spec_CA71008-000
Product specification
Customers Who Bought This Also Bought