Chloroform ≥99.8% ACS
Supplier: Avantor
Certificates
About this item
Synonyms: Methane, trichloro-,Trichloroform
Formula:CHCl₃
MW:119.38 g/mol
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
MDL Number:MFCD00000826
CAS Number:67-66-3
ADR:6.1,III
Merck Index:13,02160
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Boiling Pt:
:Boiling Pt: 61.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –63 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.38 g/mol
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
REACH:05-2114614906-45
:6.1
:III
:GHS06
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay [CHCl3 + additive(s)]:99.8% min.
- Color (APHA):10 max.
- Residue after evaporation:0.001% max.
- Acetone and aldehyde:Passes Test
- Acid and Chloride:Passes Test
- Free Chlorine (Cl):Passes Test
- Lead (Pb):0.05 ppm max.
- Substances darkened by H2SO4:Passes Test
- Presence of preservative:conforms
- Chloroform ≥99.8% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_ANAC23510-540.pdf spec_CA71006-918 spec_CA71006-922
Product specification
Customers Who Bought This Also Bought