Order Entry
ContactUsLinkComponent
Flip Glider Class Pack
Flip Glider Class Pack
Catalog #: 470211-142
Flip Glider Class Pack 35 Students
Flip Glider Class Pack
Catalog #: 470211-142

Specifications

  • Description:
    Flip Glider Class Pack
  • Cat. no.:
    470211-142
  • Supplier No.:
    502

Specifications

About this item

Features wing-tip dihedral for stability and an airfoil students can sand to shape for optimal lift.

Features wing-tip dihedral for stability and an airfoil students can sand to shape for optimal lift. Can be converted to contest version for either hand or catapult launch. Wingspan: 33.5cm (13 in), Power: Hand Launch. Grades 6 - 12. Supplies 35 students.

  • Wingspan: 33.5cm (13 in)
  • Power: Hand Launch
  • Grades 6 - 12
  • Supplies 35 students

Can be converted to contest version for either hand or catapult launch.

Customers Who Bought This Also Bought