ContactUsLinkComponent
Anaerobe Jar, Hardy Diagnostics
Anaerobe Jar, Hardy Diagnostics
Catalog #: CA89426-216
Anaerobe Jar
Anaerobe Jar, Hardy Diagnostics
Catalog #: CA89426-216
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Anaerobe Jar, Complete
  • Cat. no.:
    CA89426-216
  • Supplier No.:
    16000
Specifications
About this item
This anaerobic jar is specially formulated for temperature stability and durability
It’s unbreakable, lightweight, transparent and, of course, autoclavable. Double screw clamp, coupled with the patented Quad Ring, ensures a more positive seal, superior to conventional O-rings. It also doubles as a handy carrying handle. The 3/8” thick anodized aluminum lid won’t warp, and it’s guaranteed to resist cracking or breaking for ten years.
Delivery: Shipped complete with a stainless steel rack to accommodate 100 mm petri dishes and a stainless steel catalyst basket.
Customers Who Bought This Also Bought