Pyrite Crystal Group
Catalog #: 470210-868
Supplier: ADAM MOORE MO
Specifications
- Description:Pyrite Crystal Group
- Cat. no.:470210-868
Specifications
About this item
Choice intergrown crystal groups, many with striated pyritohedral crystal faces. Perfect for classroom study. Average size: 1" x 1". Peru.
Customers Who Bought This Also Bought