Iron Color Disc Test Kit, Model IR-21, Hach
Supplier: Avantor
Certificates
About this item
Iron color disc/TPTZ low range test kit contains 100 tests.
- Simple, fast, color disc method
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Ordering information: One reagent, two color disc, comparator box, viewing adapter, two viewing tubes, vial, instructions and carrying case included.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.