About this item
Tygon S3™ tubing is phthalate-free and is made from a NON-DEHP/bio-based plasticizer.
The sterilizing methods for this tubing are conventional autoclaving (steam 30 minutes at 15psi, 121°C) and Ethylene Oxide.
- Outstanding chemical resistance
- Consistent for reproducible results
- Easily slips over fittings and grips securely for simple lab set-ups
- Standard sizes are able to hold a full vacuum
Tygon S3™ tubing is phthalate-free and is made from a NON-DEHP/bio-based plasticizer.
This sustainable, flexible, long lasting, crack-resistant tubing continues to provide the same great performance.
This sustainable, flexible, long lasting, crack-resistant tubing continues to provide the same great performance.
The sterilizing methods for this tubing are conventional autoclaving (steam 30 minutes at 15 psi, 121 °C) and ethylene oxide.
Tygon S3™ Formulation E-3603 tubing is non-oxidizing, non-contaminating, and less permeable than rubber tubing. The glassy smooth inner bore helps prevent build-up and facilitates cleaning. Coils are also marked at 30.5 cm (1') intervals for easy measuring. It is specially formulated for resistance to flex-fatigue and abrasion. Generally used as tubing in instrumentation connection, venting, draining, and other laboratory applications, Tygon S3™ Formulation E-3603 tubing offers superior life, minimizing labor and the expense of replacement.
Specifications
- Catalog No:
- 89403-934
- 89403-936
- 89404-328
- 89404-330
- 89403-932
- 89404-340
- 89403-942
- 89404-342
- 89403-938
- 89404-336
- 89403-940
- 89404-338
- 89403-928
- 89404-326
- 89403-994
- 89403-996
- 89403-930
- 89403-992
- 89404-324
- 89404-002
- 89403-998
- 89403-846
- 89404-010
- 89403-958
- 89403-966
- 89403-960
- 89403-862
- 89404-018
- 89403-858
- 89404-012
- 89404-346
- 89404-014
- 89404-024
- 89403-870
- 89404-026
- 89404-020
- 89404-028
- 89403-972
- 89403-978
- 89403-980
- 89403-974
- 89403-976
- 89403-986
- 89403-898
- 89403-982
- 89403-984
- 89404-348
- 89404-350
- 89403-970
- 89404-310
- 89403-914
- 89404-320
- 89404-314
- 89403-916
- 89404-374
- Model:
- 89403-934
- 89403-936
- 89404-328
- 89404-330
- 89403-932
- 89404-340
- 89403-942
- 89404-342
- 89403-938
- 89404-336
- 89403-940
- 89404-338
- 89403-928
- 89404-326
- 89403-994
- 89403-996
- 89403-930
- 89403-992
- 89404-324
- 89404-002
- 89403-998
- 89403-846
- 89404-010
- 89403-958
- 89403-966
- 89403-960
- 89403-862
- 89404-018
- 89403-858
- 89404-012
- 89404-346
- 89404-014
- 89404-024
- 89403-870
- 89404-026
- 89404-020
- 89404-028
- 89403-972
- 89403-978
- 89403-980
- 89403-974
- 89403-976
- 89403-986
- 89403-898
- 89403-982
- 89403-984
- 89404-348
- 89404-350
- 89403-970
- 89404-310
- 89403-914
- 89404-320
- 89404-314
- 89403-916
- 89404-374
- Max. vacuum:
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 21.0" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 28.0" Hg @ 73 °F
- 760.0" Hg @ 73 °F
- 15.0" Hg @ 73 °F
- 760.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 12.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 7.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 13.0" Hg @ 73 °F
- 12.0" Hg @ 73 °F
- 15.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 7.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 7.0" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 6.0" Hg @ 73 °F
- 9.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 760.0" Hg @ 73 °F
- 559.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 9.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 533.0" Hg @ 73 °F
- Working pressure:
- 25.0 psi
- 25.0 psi
- 20.0 psi
- 12.0 psi
- 25.0 psi
- 47.0 psi
- 22.0 psi
- 1.7 psi
- 17.0 psi
- 2.5 psi
- 30.0 psi
- 12.0 psi
- 15.0 psi
- 40.0 psi
- 36.0 psi
- 47.0 psi
- 15.0 psi
- 12.0 psi
- 53.0 psi
- 36.0 psi
- 69.0 psi
- 36.0 psi
- 23.0 psi
- 36.0 psi
- 24.0 psi
- 69.0 psi
- 20.0 psi
- 13.0 psi
- 25.0 psi
- 36.0 psi
- 17.0 psi
- 17.0 psi
- 18.0 psi
- 40.0 psi
- 13.0 psi
- 43.0 psi
- 13.0 psi
- 20.0 psi
- 12.0 psi
- 36.0 psi
- 20.0 psi
- 25.0 psi
- 12.0 psi
- 14.0 psi
- 36.0 psi
- 20.0 psi
- 1.6 psi
- 1.4 psi
- 36.0 psi
- 20.0 psi
- 30.0 psi
- 14.0 psi
- 35.0 psi
- 10.0 psi
- 1.4 psi
Specifications
Related Information
Certificates
Search by Lot Number89403-858_24728017 89403-858_31409219 89403-862_24621669 89403-862_25179868 89403-862_25339927