About this item
Trypticase Soy Agar is a general-purpose medium for microbiology which supports the growth of fastidious as well as nonfastidious microorganisms.
- Contains casein and soy peptones for a source of general growth
- Sodium chloride maintains osmotic equilibrium
Lecithin and polysorbate 80, two commonly used neutralizers, are reported to inactivate residual disinfectants when the sample is being collected.