Kapak SilverPAK Pouches, Ampac Flexibles, A&D Weighing
Supplier: Ampac Flexibles
Certificates
About this item
Ampac's SilverPAK is a 2ply structure consisting of metalized polyester and polyethylene (METPET/PE).
These metalized pouches are ideal for powders and other granular materials.
Specifications
- Catalog No:
- 11213-921
- 11213-954
- Material:
- Silver METPET
- Silver METPET