Magnesium chloride 0.080 M, cartridge for Digital Titrator
Catalog #: CA97026-348
Supplier: Hach
CAS Number: 7786-30-3
Specifications
- Size:1 cartridge
- Cat. no.:CA97026-348
- Supplier No.:2062501
Specifications
Specification Test Results
- Method:8352
- Method Name:Magnesium Chloride
- Number of tests:40 - 100
- Parameter:Chelant, Free
- Platform :Digital Titrator
- Range:0 - 20.0 mg/L CaCO3
- Titrant:0.0800 M MgCl2
Specification Test Results
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 715 °C
:Flash Pt:
:Flash Pt:
:M.W. 95.21 g/mol
:Magnesium chloride
:715
:1412
Formula:MgCl₂
MW:95.21 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00011106
CAS Number:7786-30-3
Customers Who Bought This Also Bought