Microcentrifuge Tubes, Low Surface Tension, Graduated, with ClickLok™ MCT Seals, Simport Scientific
Supplier: Avantor
Certificates
About this item
The special type of plastic used provides these tubes with a low adhesion surface and optimum sample yield.
No lubricant (such as silicone) is necessary, thereby eliminating the danger of sample contamination. It is also graduated and designed to withstand the stress of high speed centrifugation up to 20000×g. One-piece construction with a snug-fitting attached cap and a reinforced tube bottom is added for protection against leakage.
Certifications: Certified RNase-, DNase-, Pyrogen- and DNA-free.
Ordering information: Packaged in tamperproof resealable safety-lock bags.
Customers Who Bought This Also Bought