ContactUsLinkComponent
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Catalog #: 70680-270
Covamat* Lining
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Catalog #: 70680-270
Specifications
  • Size:
    76 cm (30") wide x 15.2 meter (50 ft)
  • Cat. no.:
    70680-270
  • Supplier No.:
    246790000
Specifications
About this item
Clear polyethylene ribbed matting can be used for lining laboratory tables, shelves, trays, etc.
  • Clear polyethylene construction
  • Ribbed design
  • Easily trimmed to fit laboratory surfaces
It is easily trimmed with scissors and also works well under drums or carboys. 0.25 mm (0.010") with 0.7 mm (0.030") thick ribs.
Customers Who Bought This Also Bought