Arsenazo III indicator for metal titration, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid,2,2′-(1,8-Dihydroxy-3,6-disulfonaphthylene-2,7-bisazo)bisbenzenearsonic acid,2
Formula:C₂₂H₁₈As₂N₄O₁₄S₂
MW:776.38 g/mol
MDL Number:MFCD00036695
CAS Number:1668-00-4
ADR:6.1,
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 776.38 g/mol
EINECS:216-788-6
UN:3465
:6.1
:GHS06
:GHS09
:MQ200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:776.36 g/mol
- Bulk Density:850 kg/m3
- pH:2.3 (10 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
- Millipore®, Arsenazo III indicator for metal titration:
Specification Test Results
Related Information
Customers Who Bought This Also Bought