Iron(II) chloride tetrahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Ferrous chloride tetrahydrate
Formula:FeCl₂·4H₂O
MW:198.81 g/mol
Boiling Pt:1,023 °C
Melting Pt:105…110 °C (–H₂O of crystall.) (–H₂O of crystall.)
Density:1.936 g/cm³ (20 °C)
MDL Number:MFCD00149709
CAS Number:13478-10-9
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt: 1,023 °C
:Melting Pt:
:Melting Pt: 105…110 °C (–H₂O of crystall.) (–H₂O of crystall.)
:Flash Pt:
:Flash Pt:
:M.W. 198.81 g/mol
:Iron(II) chloride tetrahydrate
EINECS:231-843-4
:105…110 °C (–H₂O of crystall.)
:1,023 °C
Storage Temperature:Ambient
UN:3260
:GHS05
:GHS07
:MQ300
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (manganometric):≥ 98.0 %
- Sulfate (SO₄):≤ 0.01 %
- Total nitrogen (N):≤ 0.001 %
- As (Arsenic):≤ 0.0005 %
- Cu (Copper):≤ 0.002 %
- Mn (Manganese):≤ 0.005 %
- Pb (Lead):≤ 0.001 %
- Zn (Zinc):≤ 0.003 %
- Substances not precipitated by ammonia (as sulfates):≤ 0.05 %
- Supelco®, Iron(II) chloride tetrahydrate, EMSURE® for analysis:
Specification Test Results
Related Information
Customers Who Bought This Also Bought