Perchloric acid ≥70%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Formula:HClO₄
MW:100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
MDL Number:MFCD00011325
CAS Number:7601-90-3
ADR:5.1 (8),I
:Length:
:Width:
:Height:
:Ø
:Thickness:
:70
:Boiling Pt:
:Boiling Pt: 160…181 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –112.2 °C
:Flash Pt:
:Flash Pt:
:M.W. N/A N/A
Storage Temperature:Ambient
EINECS:231-512-4
:160…181 °C (1013 hPa)
UN:1873
:5.1 (8)
:I
:GHS03
:GHS05
:GHS08
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-18 °C
- Boiling Point:198.7 °C (1013 hPa)
- Density:1.68 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Solutions for Sample Digestion Flyer EMD Millipore Trace Elemental Analysis for USP 232-233 Brochure MilliporeSigma High Purity Acids and Bases
Product Literature
Ultrapur and Suprapur®High Purity Acid and Bases VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought