Evaporator Flasks, Indented, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These flasks are also referred to as drying flasks
They are particularly suited for the drying of powdered samples. The baffles, indented into the glass, provide better circulation and mixing of the powders while rotating. Plain, not coated.
Ordering information: Not for use under vacuum or pressure.
Customers Who Bought This Also Bought