Jacketed Evaporator Receiving Flask, Ace Glass Incorporated
Catalog #: 89215-046
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:8 L
- Description:Receiving Flask, Jacketed, 8 L
- Cat. no.:89215-046
- Fits Models:All R220
- Supplier No.:6727-10
Specifications
About this item
New design, jacketed receiving flask
Fits all rotary evaporators. Outer jacket has 28/15 o-ring ball joints to seal to chiller hoses. Has an SVL thread vent cap on side joint. Bottom DN25 ball joint has CAPFE o-ring. Jacket inlet and outlet ball joints have FETFE o-rings
Customers Who Bought This Also Bought