Cryovial® Workstation Rack for Cryogenic Vials, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
This handy autoclavable rack can hold up to 50 to 52 cryogenic vials.
Made of polypropylene.
Thanks to an innovative universal locking system, the vials will securely lock in each well and will not turn. Each position is identified with an alphanumeric index. Strong handles make it easy and safe to carry. It is supported by anti-skid rubber feet. The rack is compact and stackable.
Customers Who Bought This Also Bought