SP Bel-Art Silicone Rubber Loops, Bel-Art Products, a part of SP
Catalog #: 11215-974
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Silicone rubber loops
- Cat. no.:11215-974
- Supplier No.:203080000
Specifications
About this item
Use these adjustable “rubber bands” for bundling, binding, color coding, securing columns and burettes, controlling flexible tubing, keeping foil covers on canisters, and securing dialysis or biohazard bags.
- Adjustable silicone rubber construction
- Tension-control function
- Remains flexible in autoclave or freezer
They remain flexible from –46 to 121˚ C (–50 to 250 ˚F) for use in the autoclave or freezer. Small, tension-control band slides for a loop range of 5 mm (0.2") to 100 mm (4") in diameter. Includes 10 of each color, red, blue and yellow.
Customers Who Bought This Also Bought