Corning® BioCoat™ Intestinal Epithelium Differentiation Media Pack, Corning
Catalog #: CA-CB355058
Supplier: Corning
Specifications
- Description:Intestinal Epithelium Differentiation Media Pack
- Cat. no.:CA-CB355058
- Supplier No.:355058
Specifications
About this item
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
Includes 500mL Seeding Basal Medium, 500mL Differentiation Medium, and 2x0.5mL MITO+ Serum Extender.
Delivery: Pack contains seeding basal medium (500 ml), differentiation medium (500 ml), and MITO+ Serum Extender (2×0,5 ml).
Customers Who Bought This Also Bought