Serum Bottle, Polypropylene, WHEATON®, DWK Life Sciences
Catalog #: 16300-053
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Capacity:3 ml (0.1 oz.)
- Neck I.D.:7 mm
- Neck O.D.:13 mm
- Diameter ×H:17 x 38 mm
- Supplier No.:224007
Specifications
About this item
Serum bottles are designed for use with diluent solutions.
Not recommended for IV or other solutions which require a glass container.
They are autoclavable at 121 °C (250 °F) for 20 min.
Ordering information: This bottle accepts rubber stoppers and aluminum seals.
Customers Who Bought This Also Bought