SP Bel-Art Squibb Pear-Shaped Separatory Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Durable, translucent polypropylene plastic separatory funnels are resistant to most chemicals, including hydrofluoric acid.
- Chemically resistant
- Translucent
- Durable polypropylene construction
Feature a 4 mm stopcock plug of non-stick PTFE and screw cap closure. Steam autoclavable (except stopper) at 121ºC (250ºF)
Specifications
- Catalog No:
- 70690-210
- Stem ext.Ø:
- 9.5 mm (3/8")
- Stem length:
- 40 mm (1.57")