Nalgene® Labware 20-Piece Value Pack, Thermo Scientific
Catalog #: 30617-432
Supplier: Thermo Fisher Scientific
Specifications
- Description:Nalgene® Labware 20-Piece Value Pack
- Cat. no.:30617-432
- Supplier No.:1350-0001
Specifications
About this item
Nalgene* Labware Value Pack is an assortment of popular, everyday labware in a convenient pack.
Nalgene® Labware sets the standard for quality and superior performance for valuable research with the highest quality materials for dependable performance. Naglene® products are a safer alternative to glass, as they won't shatter or break. Millions of Naglene® labware products are used in laboratories world wide.
This convenient pack enables users to be ready for research right out of the box. The labware value pack makes a convenient and economical new lab starter kit, or is a great refresher pack for established labs. All items are chemically compatible with a wide range of common laboratory reagents, especially acids, bases, and alcohols.
Certifications: Graduated Cylinders Meet CFR 21, Part 177.1520 and ASTM E 1272 Class B accuracy requirements, as well as ISO Standard 6706. Beakers meet ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy.
Ordering information: Each value pack includes 6 straight-sided jars (500mL/PPCO), 4 unitary wash bottles (500mL/LDPE), 4 graduated cylinders (2–100mL, 500mL, 1L/PPCO), 5 Griffin low form beakers (50, 100, 250, 600, 1000mL/PPCO), and a blue Unwire™ half-rack for 30mm tubes.
Customers Who Bought This Also Bought