Taq DNA Polymerase with Standard Taq (Mg-free) Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
Taq DNA Polymerase is supplied with 10X Standard Taq (Mg-free) Reaction Buffer and MgCl2
10X Standard Taq (Mg-free) Reaction Buffer is detergent-free and designed to be compatible with existing assay systems.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.