ContactUsLinkComponent
SNAP-CELL® 505-Star, New England Biolabs
SNAP-CELL® 505-Star, New England Biolabs
Catalog #: CA101447-656
CAS Number:  
SNAP-Cell* 505-Star 50 nmol
SNAP-CELL® 505-Star, New England Biolabs
Catalog #: CA101447-656
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    SNAP-Cell 505-Star
  • Size:
    50 nmol
  • Cat. no.:
    CA101447-656
Specifications
About this item
SNAP-Cell® 505-Star is a photostable green fluorescent substrate that can be used to label SNAP-tag® fusion proteins inside living cells, on cell surfaces, or in vitro
This cell-permeable substrate (CP-6-505) is based on the single isomer 6-carboxyrhodamine 110 and is suitable for standard fluorescein filter sets. It has an excitation maximum at 504 nm and an emission maximum at 532 nm. This package contains 50 nmol of SNAP-Cell 505-Star substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.
Customers Who Bought This Also Bought