SNAP-Capture Magnetic Beads, New England Biolabs
Catalog #: CA101447-588
Supplier: New England Biolabs (NEB)
Specifications
- Description:SNAP-Capture Magnetic Beads
- Size:2 mL
- Cat. no.:CA101447-588
Specifications
About this item
The SNAP-Capture Magnetic Beads are used to selectively immobilize and magnetically separate a SNAP-tag® fusion protein from solution using magnetic agarose beads
These beads show a low non-specific absorption of proteins from a complex lysate, making them suitable for pull down applications. The SNAP-Capture Magnetic Beads are prepared by the coupling of SNAP-tag substrate benzylguanine with highly stable 75-150 µm superparamagnetic particles. Two ml of SNAP-Capture Magnetic Beads is sufficient to perform 25 immobilization assays using 80 µl of bead suspension per assay.
Customers Who Bought This Also Bought