Specifications
- Description:S-adenosylmethionine (SAM) (32mM)
- Size:0.5 mL
- Cat. no.:CA101227-496
Specifications
About this item
S-adenosyl-methionine (SAM) is included with all of the DNA methylases available from New England Biolabs, Inc
As well as certain restriction enzymes for which it is required. The SAM is prepared in a solution of 0.005 M H2SO4 and 10% ETOH and filtered.