Specifications
- Description:Quick Blunting and Quick Ligation Kits
- Cat. no.:CA101417-934
- No. of Reactions:20
Specifications
About this item
NEB offers the Quick Blunting™ and Quick Ligation™ Kits together as a bundle at a value price.
The Quick Blunting™ Kit is used to convert DNA with incompatible 5´or 3´overhangs to 5´phosphorylated, blunt-ended DNA for efficient blunt-end ligation into DNA cloning vectors. DNA is blunted using T4 DNA polymerase which has both 3´→ 5´ exonuclease activity and 5´→ 3´ polymerase activity. T4 Polynucleotide Kinase is included in the enzyme mix for phosphorylation of the 5´ends of blunt-ended DNA for subsequent ligation into a cloning vector. This kit is optimized for blunting up to 5 µg of DNA in a single reaction. The Quick Ligation™ Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature (25°C).