pTXB1 Vector DNA, New England Biolabs
Catalog #: CA101227-160
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:pTXB1 Vector DNA
- Size:10 µg
- Cat. no.:CA101227-160
- Vector Details:pTXB1
Specifications
About this item
pTXB1 is an E.coli expression vector in the IMPACT™ Kit.
It is designed for the in-frame insertion of a target gene into the polylinker upstream of the Mxe intein/chitin binding domain (27 kDa). The fusion protein is bound to chitin beads and the thiol-induced cleavage activity of the intein releases the target protein. pTXB vectors are recommended for use in intein-mediated protein ligation and C-terminal labeling. This double stranded vector is 6,706 base pairs in length.
Customers Who Bought This Also Bought